Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181392.3 | NP_852057.2 | 3 | 617 | ||
GenBank mRNA | AK003579.1 | - | 3 | 600 | ||
AK011281.1 | - | 3 | 602 | |||
AK012257.1 | - | 3 | 679 | |||
AK013225.1 | - | 3 | 581 |