Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177613.2 | NP_808281.1 | 5 | 1127 | ||
GenBank mRNA | AK136078.1 | - | 1097 | |||
AK153982.1 | - | 1 | 1916 | |||
BC039160.1 | - | 5 | 1127 | 4024472 | ||
BC094502.1 | - | 5 | 943 | 5034261 |