Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007806.3 | NP_031832.2 | 3, 4 | 279 | ||
GenBank mRNA | AK021200.1 | - | 3, 4 | 267 | ||
AK078854.1 | - | 3, 4 | 250 | |||
AK131580.1 | - | 3, 4 | 248 | |||
AK152414.1 | - | 3, 4 | 270 | |||
AK171748.1 | - | 3, 4 | 248 | |||
BC026791.1 | - | 3, 4 | 230 | 3980538 | ||
BC060635.1 | - | 2, 3 | 4285 | |||
BY005999.1 | - | 3, 4 | 282 | |||
CF738398.1 | - | 3, 4 | 262 | |||
EU791539.1 | - | 3, 4 | 209 | |||
M31775.1 | - | 3, 4 | 202 |