Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007969.4 | NP_031995.3 | 2 | 241 | ||
GenBank mRNA | BC010986.1 | - | 2 | 241 | 4038377 | |
BE310436.1 | - | 2 | 102 | |||
EH099462.1 | - | 2 | 268 | |||
EH105860.1 | - | 2 | 268 | |||
EH109450.1 | - | 2 | 147 | |||
X93037.1 | - | 2 | 139 |