Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3f3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008210.4 | NP_032236.1 | 2 | 275 | ||
GenBank mRNA | AK008405.1 | - | 1 | 175 | ||
AK037900.1 | - | 2 | 277 | |||
AK150467.1 | - | 2 | 235 | |||
AK150468.1 | - | 2 | 235 | |||
AK150591.1 | - | 2 | 230 | |||
AK150928.1 | - | 2 | 230 | |||
AK151634.1 | - | 2 | 235 | |||
AK152293.1 | - | 2 | 230 | |||
AK153034.1 | - | 2 | 234 | |||
AK160635.1 | - | 2 | 230 | |||
AK160677.1 | - | 2 | 234 | |||
AK162040.1 | - | 2 | 233 | |||
AK167879.1 | - | 2 | 234 | |||
AK168197.1 | - | 2 | 234 | |||
AK169608.1 | - | 2 | 230 | |||
AK177127.1 | - | 1 | 45 | |||
AK178894.1 | - | 1 | 45 | |||
AK180970.1 | - | 1 | 45 | |||
AK186827.1 | - | 1 | 45 | |||
AK189335.1 | - | 1 | 84 | |||
AK215513.1 | - | 1 | 45 | |||
AK216872.1 | - | 366 | ||||
AK217933.1 | - | 1 | 45 | |||
BC002268.1 | - | 2 | 220 | 3495830 | ||
BC012687.1 | - | 2 | 224 | 4224280 | ||
BC083353.1 | - | 2 | 215 | 5714859 | ||
BC088835.1 | - | 2 | 212 | 6437687 | ||
BC106177.1 | - | 2 | 209 | 6511787 | ||
Z85979.1 | - | 2 | 223 |
Gene Symbol: Gm12657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081019.1 | NP_001074488.1 | 2 | 162 |