Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303265.1 | NP_001290194.1 | 5 | 529 | ||
NM_001545.2 | NP_001536.1 | 5 | 529 | |||
XM_017024574.1 | XP_016880063.1 | 5 | 522 | |||
GenBank mRNA | AK222553.1 | - | 5 | 519 | ||
AK314138.1 | - | 5 | 517 | |||
BC015335.1 | - | 5 | 512 | 4418983 | ||
BI870683.1 | - | 5 | 386 | |||
BT007111.1 | - | 5 | 506 | |||
X81788.1 | - | 5 | 508 |