Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008843.3 | NP_032869.2 | 2, 3 | 229 | ||
GenBank mRNA | AB017907.1 | - | 2, 3 | 207 | ||
AK138795.1 | - | 2, 3 | 230 | |||
BC028889.1 | - | 2, 3 | 227 | 4165968 | ||
S73282.1 | - | 2, 3 | 225 | |||
X02510.1 | - | 2, 3 | 224 |