Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008993.2 | NP_033019.2 | 1, 2 | 283 | ||
GenBank mRNA | AF309644.1 | - | 1, 2 | 137 | ||
AK003118.1 | - | 1, 2 | 137 | |||
BC057975.1 | - | 1, 2 | 124 | 5055267 | ||
BY090564.1 | - | 1, 2 | 283 |