Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010507.1 | NP_034637.1 | 1 | 113 | ||
GenBank mRNA | BC117775.1 | - | 1 | 108 |
Gene Symbol: Ifna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC117774.1 | - | 113 | 40091177 |