Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010822.3 | NP_034952.2 | 1 | 125 | ||
GenBank mRNA | AK018694.1 | - | 1 | 105 | ||
AK076201.1 | - | 1 | 120 | |||
AK088878.1 | - | 1 | 79 | |||
AK134900.1 | - | 1 | 92 | |||
AK155766.1 | - | 1 | 126 | |||
AK156067.1 | - | 1 | 92 | |||
BC014754.1 | - | 1 | 119 | 4486938 | ||
BY309585.1 | - | 1 | 125 | |||
U10420.1 | - | 1 | 118 |