Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011664.3 | NP_035794.1 | 2 | 178 | ||
GenBank mRNA | AK003190.1 | - | 2 | 179 | ||
AK007273.1 | - | 2 | 210 | |||
AK010413.1 | - | 2 | 180 | |||
AK010625.1 | - | 2 | 179 | |||
AK012443.1 | - | 2 | 180 | |||
AK013027.1 | - | 2 | 179 | |||
AK013873.1 | - | 2 | 177 | |||
AK132747.1 | - | 2 | 179 | |||
AK160683.1 | - | 2 | 176 | |||
AK165313.1 | - | 2 | 179 | |||
BC019850.1 | - | 2 | 147 | 5123849 | ||
BC066197.1 | - | 2 | 208 | 6435236 | ||
BC100341.1 | - | 2 | 133 | 30787432 | ||
X51703.1 | - | 2 | 184 |
Gene Symbol: Gm1821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088453.1 | - | 1 | 180 |