Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011971.4 | NP_036101.1 | 2 | 182 | ||
GenBank mRNA | AF060092.1 | - | 2 | 144 | ||
AK002331.1 | - | 2 | 183 | |||
AK010488.1 | - | 2 | 163 | |||
AK050108.1 | - | 2 | 180 | |||
AK153162.1 | - | 2 | 215 | |||
BC014783.1 | - | 2 | 149 | 3500033 | ||
BC098233.1 | - | 1 | 144 | 1547611 |