Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301284.1 | NP_001288213.1 | 4, 5 | 350 | ||
NM_007806.3 | NP_031832.2 | 5, 6 | 434 | |||
GenBank mRNA | AK018713.1 | - | 4, 5 | 319 | ||
AK021200.1 | - | 5, 6 | 422 | |||
AK131580.1 | - | 5, 6 | 403 | |||
AK152414.1 | - | 5, 6 | 425 | |||
AK171748.1 | - | 5, 6 | 403 | |||
BC026791.1 | - | 5, 6 | 385 | 3980538 | ||
CF738398.1 | - | 5, 6 | 417 | |||
EU791539.1 | - | 5, 6 | 364 | |||
M31775.1 | - | 5, 6 | 357 |