Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001552.2 | NP_001543.2 | 4 | 1841 | ||
GenBank mRNA | BC016041.1 | - | 4 | 1651 | 4661581 | |
M62403.1 | - | 4 | 1612 |