Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008776.2 | NP_032802.1 | 3 | 248 | ||
GenBank mRNA | AK039375.1 | - | 3 | 231 | ||
AK154547.1 | - | 3 | 244 | |||
AK167496.1 | - | 3 | 216 | |||
BC067015.1 | - | 3 | 219 | 6820858 | ||
U57746.1 | - | 3 | 246 |