Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129803.1 | NP_001123275.1 | 3 | 219 | ||
NM_001129804.1 | NP_001123276.1 | 3 | 191 | |||
NM_008790.2 | NP_032816.1 | 3 | 216 | |||
GenBank mRNA | AK003002.1 | - | 3 | 162 | ||
AK160398.1 | - | 3 | 180 | |||
BC014694.1 | - | 2 | 338 | |||
BC024853.1 | - | 2 | 359 | |||
BC028982.1 | - | 3 | 199 | |||
BU531373.1 | - | 3 | 217 | |||
CB522989.1 | - | 3 | 191 | |||
EU164759.1 | - | 3 | 219 | |||
M21532.1 | - | 3 | 186 |