Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008838.1 | NP_032864.1 | 2 | 239 | ||
GenBank mRNA | AK132796.1 | - | 2 | 245 | ||
AK171083.1 | - | 2 | 227 | |||
AK218842.1 | - | 2 | 151 | |||
BC028862.1 | - | 2 | 228 | 3673399 | ||
D50264.1 | - | 2 | 239 |