Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009009.4 | NP_033035.3 | 3 | 482 | ||
GenBank mRNA | AF332085.1 | - | 2 | 237 | ||
AF332086.1 | - | 2 | 237 | |||
AK004746.1 | - | 3 | 285 | |||
AK128984.1 | - | 3 | 371 | |||
AK139492.1 | - | 2 | 135 | |||
AK145591.1 | - | 3 | 449 | |||
AK163992.1 | - | 343 | ||||
AK168718.1 | - | 3 | 470 | |||
AK168896.1 | - | 3 | 431 | |||
BC043032.1 | - | 2 | 251 | 5696540 | ||
BC129916.1 | - | 2 | 281 | 40111820 | ||
BC129917.1 | - | 2 | 282 | 40111821 | ||
X98293.1 | - | 2 | 237 |