Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009034.4 | NP_033060.3 | 2 | 234 | ||
GenBank mRNA | AK003312.1 | - | 2 | 219 | ||
BC106846.2 | - | 2 | 243 | |||
BC125551.1 | - | 2 | 203 | 40130066 | ||
BC125553.1 | - | 2 | 203 | 40130068 | ||
X74154.1 | - | 2 | 218 |