Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011409.1 | NP_035539.1 | 2 | 342 | ||
GenBank mRNA | AF099974.1 | - | 2 | 342 | ||
AK133208.1 | - | 2 | 1694 | |||
BC132627.1 | - | 2 | 280 | 40130904 | ||
BC138070.1 | - | 2 | 280 | 8861089 | ||
BC145134.1 | - | 3 | 447 | 9053682 |
Gene Symbol: Slfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137559.1 | - | 7 | 1618 | ||
AK137689.1 | - | 6 | 1539 | |||
BC044865.1 | - | 7 | 1627 | 4459564 | ||
BC150770.1 | - | 4 | 1234 | 9087681 |