Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011971.4 | NP_036101.1 | 3 | 266 | ||
GenBank mRNA | AF060092.1 | - | 3 | 228 | ||
AK002331.1 | - | 3 | 267 | |||
AK010488.1 | - | 3 | 247 | |||
AK050108.1 | - | 3 | 264 | |||
AK080755.1 | - | 3 | 266 | |||
AK153162.1 | - | 3 | 299 | |||
BC014783.1 | - | 3 | 233 | 3500033 | ||
BC098233.1 | - | 2 | 228 | 1547611 |