Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013560.2 | NP_038588.2 | 1 | 484 | ||
GenBank mRNA | AF047377.1 | - | 2 | 177 | ||
AF047378.1 | - | 2 | 101 | |||
AK003119.1 | - | 1 | 405 | |||
AK010663.1 | - | 1 | 402 | |||
AK168927.1 | - | 1 | 402 | |||
BC018257.1 | - | 1 | 424 | 3498180 | ||
BC090388.1 | - | 1 | 385 | |||
BC099463.1 | - | 1 | 379 | 30848584 | ||
U03560.1 | - | 1 | 341 | |||
U03561.1 | - | 2 | 308 | |||
U03562.1 | - | 2 | 242 | |||
X14686.1 | - | 1 | 386 | |||
X14687.1 | - | 1 | 431 |