Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcl1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013772.2 | NP_038800.1 | 1 | 176 | ||
GenBank mRNA | AF195490.1 | - | 1 | 178 | ||
BC119260.1 | - | 1 | 176 | 8734009 | ||
BC120575.1 | - | 1 | 176 | 40129498 |