Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013895.4 | NP_038923.1 | 1 | 306 | ||
GenBank mRNA | AF144702.1 | - | 1 | 308 | ||
AK003054.1 | - | 1 | 112 | |||
AK009041.1 | - | 1 | 100 | |||
AK009152.1 | - | 1 | 116 | |||
AK013230.1 | - | 1 | 104 | |||
AK088984.1 | - | 1 | 116 | |||
AK176965.1 | - | 1 | 25 | |||
BC011436.1 | - | 1 | 119 | 2812494 |