Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022422.5 | NP_071867.1 | 2 | 217 | ||
GenBank mRNA | AB030194.1 | - | 2 | 166 | ||
AK018778.1 | - | 2 | 166 | |||
AY029485.1 | - | 2 | 166 | |||
BC048431.1 | - | 2 | 165 | 6601662 | ||
BC059712.1 | - | 2 | 164 | 6544762 | ||
BU554843.1 | - | 2 | 221 | |||
EH103048.1 | - | 44 |