Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022888.2 | NP_075026.1 | 2 | 352 | ||
NM_176807.4 | NP_789777.1 | 2 | 352 | |||
GenBank mRNA | AF250145.1 | - | 2 | 352 | ||
AK003087.1 | - | 1 | 159 | |||
AK041596.1 | - | 2 | 286 | |||
AK083306.1 | - | 2 | 353 | |||
BC028431.1 | - | 2 | 333 | 1265235 | ||
EU326437.1 | - | 1 | 110 | |||
EU326438.1 | - | 1 | 110 | |||
EU326439.1 | - | 1 | 110 | |||
EU326440.1 | - | 1 | 110 | |||
EU326441.1 | - | 1 | 110 | |||
EU326442.1 | - | 1 | 110 |