Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014383.2 | - | 2 | 473 | ||
AK045045.1 | - | 2 | 607 | |||
BC103778.1 | - | 2 | 541 | 6493935 | ||
BC116935.1 | - | 2 | 459 | 40126254 | ||
BC116937.1 | - | 2 | 461 | 40126256 |