Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcl1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013775.1 | NP_038803.1 | 3, 4 | 416 | ||
GenBank mRNA | AF195489.1 | - | 3, 4 | 416 | ||
BC099461.1 | - | 3, 4 | 400 | 30935073 |