Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282061.1 | NP_001268990.1 | 3 | 232 | ||
NM_015812.3 | NP_056627.1 | 3 | 514 | |||
GenBank mRNA | AK052527.1 | - | 3 | 515 | ||
AK053087.1 | - | 3 | 233 | |||
AK139323.1 | - | 3 | 629 | |||
BC038616.1 | - | 1 | 59 | |||
BC125029.1 | - | 3 | 218 | 40062355 | ||
BC125030.1 | - | 3 | 218 | 40062356 |