Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159616.1 | NP_001153088.1 | 4 | 332 | ||
NM_001159617.1 | NP_001153089.1 | 4 | 501 | |||
NM_001159618.1 | NP_001153090.1 | 4 | 411 | |||
NM_001159619.1 | NP_001153091.1 | 3 | 364 | |||
NM_001159620.1 | NP_001153092.1 | 3 | 364 | |||
NM_019543.3 | NP_062416.1 | 4 | 347 | |||
GenBank mRNA | AF216306.1 | - | 4 | 334 | ||
AF216307.1 | - | 4 | 309 | |||
AF216308.1 | - | 3 | 254 | |||
AF216309.1 | - | 4 | 312 | |||
AK004169.1 | - | 4 | 311 | |||
BC038252.1 | - | 4 | 512 | 4913346 | ||
BC061116.1 | - | 4 | 285 | 6593482 | ||
BC061117.1 | - | 3 | 323 | |||
BC096569.1 | - | 4 | 339 | 1429742 | ||
BX511452.1 | - | 4 | 412 | |||
BY041400.1 | - | 4 | 332 | |||
CB596599.1 | - | 3 | 367 |