Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cited4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019563.2 | NP_062509.1 | 1 | 736 | ||
GenBank mRNA | AF143369.1 | - | 1 | 706 | ||
AK012136.1 | - | 1 | 728 | |||
AK154030.1 | - | 1 | 727 | |||
BC025116.1 | - | 2 | 408 | |||
BC080723.1 | - | 2 | 493 | |||
BC132360.1 | - | 1 | 683 | 40130637 | ||
BC132362.1 | - | 1 | 683 | 40130639 |