Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019990.4 | NP_064374.1 | 2 | 335 | ||
GenBank mRNA | AB031550.1 | - | 3 | 512 | ||
BC046335.1 | - | 2 | 424 | |||
BC058773.1 | - | 2 | 467 | |||
BC080808.1 | - | 2 | 530 | |||
BC094007.1 | - | 2 | 348 | |||
BC116984.1 | - | 1 | 243 | 40126303 | ||
BC120641.1 | - | 1 | 243 | 40129564 | ||
BC144877.1 | - | 1 | 243 | |||
BG078855.2 | - | 1 | 121 | |||
BY079536.1 | - | 2 | 341 |