Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025417.2 | NP_079693.1 | 4 | 180 | ||
GenBank mRNA | AK004174.1 | - | 4 | 181 | ||
AK009802.1 | - | 4 | 179 | |||
AK172194.1 | - | 3 | 1292 | |||
BC033438.1 | - | 3 | 166 | 3995599 | ||
BC049222.1 | - | 4 | 179 | 2649359 | ||
BC055798.1 | - | 3 | 1271 | |||
DQ702594.1 | - | 11 |