Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082532.1 | NP_001076001.1 | 2 | 301 | ||
GenBank mRNA | AK007367.1 | - | 2 | 282 | ||
AK021110.1 | - | 2 | 282 | |||
BC025114.1 | - | 2 | 261 | |||
BU558514.1 | - | 2 | 304 |