Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam122a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026520.3 | NP_080796.1 | 1 | 476 | ||
GenBank mRNA | AK005219.1 | - | 1 | 466 | ||
AK010777.1 | - | 1 | 487 | |||
AK011741.1 | - | 1 | 460 | |||
AK018219.1 | - | 1 | 431 | |||
AK033646.1 | - | 1 | 476 | |||
AK077654.1 | - | 1 | 458 | |||
AK172089.1 | - | 1 | 461 | |||
BC028637.1 | - | 1 | 444 | 1248203 | ||
BC083062.1 | - | 1 | 466 | 5361132 |