Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310067B10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028014.3 | NP_082290.2 | 32 | 4668 | ||
GenBank mRNA | AK010082.1 | - | 3 | 667 | ||
AK047622.1 | - | 18 | 2802 | |||
BC007157.1 | - | 19 | 2914 | |||
BC030865.1 | - | 17 | 2435 | |||
BC053088.1 | - | 32 | 4583 | 5709104 | ||
CA325552.1 | - | 1 | 165 |