Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029565.3 | NP_083841.4 | 6 | 756 | ||
GenBank mRNA | AK003252.1 | - | 6 | 670 | ||
AK014184.1 | - | 6 | 542 | |||
AK050162.1 | - | 6 | 674 | |||
AK145644.1 | - | 6 | 677 | |||
AK159761.1 | - | 6 | 684 | |||
AK166772.1 | - | 6 | 671 | |||
AK167058.1 | - | 6 | 670 | |||
AK177231.1 | - | 2 | 89 | |||
AX775905.1 | - | 6 | 691 | |||
BC014732.1 | - | 6 | 660 | 4235248 | ||
BC018379.1 | - | 6 | 678 | 4242583 | ||
BC045145.1 | - | 6 | 694 | 6475115 | ||
BC058273.1 | - | 6 | 671 | 6494379 |