Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031248.3 | NP_112538.1 | 2 | 271 | ||
GenBank mRNA | AJ277386.1 | - | 2 | 241 | ||
AK008126.1 | - | 2 | 288 | |||
AK008185.1 | - | 2 | 291 | |||
AK008395.1 | - | 2 | 288 | |||
AK019006.1 | - | 2 | 266 | |||
BC031816.1 | - | 2 | 271 | 4189054 |