Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700020L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025492.3 | NP_079768.2 | 3 | 518 | ||
GenBank mRNA | AK005809.1 | - | 3 | 517 | ||
AK006174.1 | - | 3 | 519 | |||
BC152320.1 | - | 3 | 505 | 40136395 |