Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026020.6 | NP_080296.3 | 3 | 214 | ||
GenBank mRNA | AK002419.1 | - | 3 | 181 | ||
AK008344.1 | - | 3 | 215 | |||
AK010618.1 | - | 3 | 221 | |||
AK012402.1 | - | 3 | 209 | |||
AK028140.1 | - | 3 | 213 | |||
AK028149.1 | - | 3 | 215 | |||
AK088737.1 | - | 3 | 214 | |||
AK134206.1 | - | 3 | 214 | |||
AK166774.1 | - | 3 | 179 | |||
BC012413.1 | - | 3 | 196 | 3583752 |