Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026066.2 | NP_080342.1 | 1 | 277 | ||
NM_029390.1 | NP_083666.1 | 1 | 250 | |||
GenBank mRNA | AK005168.1 | - | 1 | 284 | ||
AK005375.1 | - | 1 | 232 | |||
AK013666.1 | - | 1 | 277 | |||
AY241867.1 | - | 1 | 267 | |||
BC049665.1 | - | 1 | 264 | 6533928 | ||
DQ115983.1 | - | 1 | 250 |