Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam122a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026520.3 | NP_080796.1 | 1 | 488 | ||
GenBank mRNA | AK005219.1 | - | 1 | 478 | ||
AK010777.1 | - | 1 | 499 | |||
AK011741.1 | - | 1 | 472 | |||
AK033646.1 | - | 1 | 488 | |||
AK077654.1 | - | 1 | 470 | |||
AK172089.1 | - | 1 | 473 | |||
BC028637.1 | - | 1 | 456 | 1248203 | ||
BC083062.1 | - | 1 | 478 | 5361132 |