Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027215.2 | NP_081491.2 | 4 | 416 | ||
GenBank mRNA | AK007753.1 | - | 4 | 377 | ||
AK008089.1 | - | 4 | 370 | |||
AK010168.1 | - | 4 | 405 | |||
AK160267.1 | - | 4 | 419 | |||
BC147300.1 | - | 4 | 370 | 9056333 | ||
BC147301.1 | - | 4 | 370 | 9056336 |