Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321065.1 | NP_001307994.1 | 3 | 385 | ||
NM_001321066.1 | NP_001307995.1 | 4 | 447 | |||
NM_001645.4 | NP_001636.1 | 3 | 321 | |||
GenBank mRNA | AK225971.1 | - | 3 | 307 | ||
AK312036.1 | - | 3 | 202 | |||
BC009698.2 | - | 2 | 144 | 3889900 | ||
BC055093.1 | - | 4 | 373 | 6567705 | ||
BT007142.1 | - | 2 | 122 | |||
CR456907.1 | - | 2 | 122 | |||
DA482306.1 | - | 4 | 447 | |||
M27359.1 | - | 2 | 80 | |||
X00570.1 | - | 2 | 178 |