Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029565.3 | NP_083841.4 | 7, 8 | 923 | ||
GenBank mRNA | AK003252.1 | - | 7, 8 | 837 | ||
AK014184.1 | - | 7, 8 | 709 | |||
AK050162.1 | - | 7, 8 | 841 | |||
AK145644.1 | - | 7, 8 | 844 | |||
AK159761.1 | - | 7, 8 | 851 | |||
AK166772.1 | - | 7, 8 | 838 | |||
AK167058.1 | - | 7, 8 | 837 | |||
AK177231.1 | - | 3, 4 | 256 | |||
AX775905.1 | - | 7, 8 | 858 | |||
BC002164.1 | - | 1, 2 | 102 | |||
BC014732.1 | - | 7, 8 | 827 | 4235248 | ||
BC018379.1 | - | 7, 8 | 845 | 4242583 | ||
BC045145.1 | - | 7, 8 | 861 | 6475115 | ||
BC058273.1 | - | 7, 8 | 838 | 6494379 |