Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030694.1 | NP_109619.1 | 2 | 315 | ||
GenBank mRNA | AK141392.1 | - | 2 | 342 | ||
AK167865.1 | - | 2 | 53 | |||
AY082486.1 | - | 2 | 337 | |||
BC002102.1 | - | 2 | 315 | 3485116 | ||
BC002160.1 | - | 2 | 320 | 3485614 | ||
BC084679.1 | - | 2 | 315 | 5374182 |