Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007503.3 | NP_031529.2 | 2, 3 | 121 | ||
NM_052823.2 | NP_439888.1 | 2, 3 | 194 | |||
GenBank mRNA | AA396010.1 | - | 2, 3 | 142 | ||
AK002215.1 | - | 2, 3 | 109 | |||
AK002228.1 | - | 2, 3 | 122 | |||
AY626243.1 | - | 2, 3 | 243 | |||
AY626244.1 | - | 2, 3 | 194 | |||
AY626245.1 | - | 2, 3 | 279 | |||
BC024614.1 | - | 2, 3 | 110 | 4222609 | ||
BC069892.1 | - | 2, 3 | 118 | |||
CA491597.1 | - | 2, 3 | 360 | |||
CB600439.1 | - | 2, 3 | 154 | |||
CB600958.1 | - | 2, 3 | 329 | |||
X70060.1 | - | 1, 2 | 32 |