Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053191.2 | NP_444421.2 | 2 | 377 | ||
GenBank mRNA | AF329196.1 | - | 2 | 297 | ||
AK030960.1 | - | 2 | 378 | |||
AK040961.1 | - | 2 | 376 | |||
AK165584.1 | - | 1 | 53 | |||
BC116705.1 | - | 2 | 273 | 40102877 |