Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027179.1 | NP_081455.1 | 3 | 244 | ||
NM_134118.5 | NP_598879.1 | 3 | 244 | |||
GenBank mRNA | AK002384.1 | - | 3 | 215 | ||
AK010513.1 | - | 3 | 180 | |||
AK010984.1 | - | 3 | 218 | |||
AK013558.1 | - | 3 | 200 | |||
AK020708.1 | - | 3 | 238 | |||
BC019984.1 | - | 3 | 230 | 4240045 | ||
BC094046.1 | - | 3 | 182 | 5064002 | ||
BY278351.1 | - | 3 | 244 |