Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001662.3 | NP_001653.1 | 6 | 642 | ||
GenBank mRNA | AF493884.1 | - | 6 | 495 | ||
AK308548.1 | - | 5 | 1295 | |||
AK312107.1 | - | 6 | 547 | |||
BC003043.1 | - | 6 | 532 | 2822973 | ||
BC033104.1 | - | 6 | 589 | 4862014 | ||
BT007087.1 | - | 6 | 495 | |||
M57567.1 | - | 6 | 549 |